Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLP3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126039.4 | NP_569033.1 | 7 - 8 | 1056 | 106 | |
GenBank mRNA | AY086393.1 | - | 7 - 8 | 687 | 106 | |
BT003998.1 | - | 7 - 8 | 723 | 106 | ||
BT005683.1 | - | 7 - 8 | 584 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |