Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126066.4 | NP_569038.1 | 5 - 6 | 824 | 67 | |
GenBank mRNA | AF428338.1 | - | 5 - 6 | 742 | 67 | |
AY150381.1 | - | 5 - 6 | 701 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |