Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKKK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345806.1 | NP_001330515.1 | 9 - 10 | 2163 | 96 | |
NM_126084.3 | NP_569040.1 | 9 - 10 | 2163 | 96 | ||
GenBank mRNA | AF360242.1 | - | 9 - 10 | 2061 | 96 | |
AY040040.1 | - | 9 - 10 | 1896 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |