Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126091.4 | NP_569041.1 | 2 - 3 | 540 | 85 | |
GenBank mRNA | AY094401.1 | - | 2 - 3 | 391 | 85 | |
BT000862.1 | - | 2 - 3 | 278 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |