Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345819.1 | NP_001331563.1 | 12 - 13 | 1706 | 104 | |
NM_126124.4 | NP_201525.2 | 12 - 13 | 1818 | 104 | ||
GenBank mRNA | AY080843.1 | - | 12 - 13 | 1669 | 104 | |
BT001977.1 | - | 12 - 13 | 1598 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |