Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345817.1 | NP_001318895.1 | 3 - 4 | 414 | 100 | |
NM_001345818.1 | NP_001331564.1 | 3 - 4 | 438 | 100 | ||
NM_001345819.1 | NP_001331563.1 | 3 - 4 | 414 | 100 | ||
NM_126124.4 | NP_201525.2 | 3 - 4 | 526 | 100 | ||
GenBank mRNA | AY080843.1 | - | 3 - 4 | 377 | 100 | |
BT001977.1 | - | 3 - 4 | 306 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |