Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126132.4 | NP_201533.1 | 5 - 6 | 957 | 59 | |
GenBank mRNA | AY057698.1 | - | 5 - 6 | 957 | 59 | |
AY143932.1 | - | 5 - 6 | 843 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |