Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345832.1 | NP_001330470.1 | 2 - 3 | 500 | 108 | |
NM_126148.4 | NP_201549.1 | 2 - 3 | 746 | 108 | ||
NM_203278.2 | NP_975007.1 | 1 - 2 | 564 | 108 | ||
GenBank mRNA | AF446873.1 | - | 1 - 2 | 235 | 108 | |
AY052213.1 | - | 2 - 3 | 384 | 108 | ||
AY316677.1 | - | 1 - 2 | 235 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |