Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QSOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126188.4 | NP_565258.1 | 11 - 12 | 1617 | 113 | |
GenBank mRNA | AY090364.1 | - | 11 - 12 | 1591 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |