Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126189.4 | NP_178237.3 | 15 - 16 | 1406 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |