Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335052.1 | NP_001325195.1 | 4 - 5 | 295 | 59 | |
NM_001335053.1 | NP_001325196.1 | 4 - 5 | 295 | 59 | ||
NM_126189.4 | NP_178237.3 | 4 - 5 | 295 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |