Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126204.3 | NP_178252.2 | 3 - 4 | 567 | 78 | |
GenBank mRNA | AJ431181.1 | - | 3 - 4 | 567 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Arabidopsis thaliana Probe spans exons |