Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035862.2 | NP_001030939.1 | 7 - 8 | 1120 | 115 | |
NM_001035863.1 | NP_001030940.1 | 7 - 8 | 1202 | 115 | ||
NM_001035864.1 | NP_001030941.1 | 7 - 8 | 1147 | 115 | ||
NM_126206.4 | NP_178254.2 | 7 - 8 | 1222 | 115 | ||
GenBank mRNA | AK228266.1 | - | 7 - 8 | 1113 | 115 | |
AK317182.1 | - | 7 - 8 | 1120 | 115 | ||
BT006469.1 | - | 6 - 7 | 1036 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |