Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335070.1 | NP_001318177.1 | 1 - 2 | 982 | 107 | |
NM_001335071.1 | NP_001323562.1 | 1 - 2 | 982 | 107 | ||
NM_126226.4 | NP_565271.1 | 1 - 2 | 996 | 107 | ||
GenBank mRNA | AK118911.1 | - | 1 - 2 | 952 | 107 | |
AY572782.1 | - | 1 - 2 | 861 | 107 | ||
BT005408.1 | - | 1 - 2 | 861 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |