Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335070.1 | NP_001318177.1 | 2 - 3 | 1270 | 72 | |
NM_126226.4 | NP_565271.1 | 2 - 3 | 1284 | 72 | ||
GenBank mRNA | AK118911.1 | - | 2 - 3 | 1240 | 72 | |
AY572782.1 | - | 2 - 3 | 1149 | 72 | ||
BT005408.1 | - | 2 - 3 | 1149 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |