Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPSF73-II | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126234.3 | NP_178282.2 | 2 - 3 | 368 | 75 | |
GenBank mRNA | AK221561.1 | - | 2 - 3 | 300 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |