Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP70-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202568.1 | NP_001189497.1 | 5 - 6 | 758 | 72 | |
NM_126236.4 | NP_178284.1 | 5 - 6 | 822 | 72 | ||
GenBank mRNA | AM086440.1 | - | 4 - 5 | 577 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |