Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126238.3 | NP_178286.1 | 2 - 3 | 379 | 62 | |
GenBank mRNA | AY316134.1 | - | 2 - 3 | 355 | 62 | |
BT028987.1 | - | 2 - 3 | 342 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |