Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126249.5 | NP_178297.2 | 2 - 3 | 567 | 137 | |
GenBank mRNA | AF492659.1 | - | 2 - 3 | 350 | 137 | |
BT026449.1 | - | 2 - 3 | 350 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |