Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126249.5 | NP_178297.2 | 4 - 5 | 800 | 150 | |
GenBank mRNA | AF492659.1 | - | 4 - 5 | 583 | 150 | |
BT026449.1 | - | 4 - 5 | 583 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |