Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-B5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126288.3 | NP_178336.1 | 1 - 2 | 432 | 69 | |
GenBank mRNA | AY219138.1 | - | 1 - 2 | 353 | 69 | |
BT026045.1 | - | 1 - 2 | 338 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |