Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126290.2 | NP_178338.1 | 2 - 3 | 495 | 84 | |
GenBank mRNA | AY649301.1 | - | 2 - 3 | 472 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |