Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PP2-B8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126291.2 | NP_178339.1 | 2 - 3 | 523 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |