Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335105.1 | NP_001318181.1 | 1 - 2 | 580 | 110 | |
NM_126292.3 | NP_565282.3 | 2 - 3 | 452 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |