Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHY3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126329.4 | NP_178377.2 | 3 - 4 | 480 | 87 | |
GenBank mRNA | BT015336.1 | - | 3 - 4 | 433 | 87 | |
BT015641.1 | - | 3 - 4 | 425 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |