Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126336.3 | NP_565290.1 | 1 - 2 | 221 | 67 | |
GenBank mRNA | AF370496.1 | - | 1 - 2 | 220 | 67 | |
BT020431.1 | - | 1 - 2 | 105 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |