Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126363.7 | NP_565294.1 | 6 - 7 | 749 | 103 | |
GenBank mRNA | AY065169.1 | - | 6 - 7 | 711 | 103 | |
AY114571.1 | - | 6 - 7 | 578 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |