Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035542.1 | NP_001030619.1 | 4 - 5 | 728 | 64 | |
NM_111020.3 | NP_566141.1 | 4 - 5 | 771 | 64 | ||
NM_180161.2 | NP_850492.1 | 4 - 5 | 793 | 64 | ||
NM_202477.2 | NP_974206.1 | 4 - 5 | 720 | 64 | ||
GenBank mRNA | AB010259.1 | - | 3 - 4 | 526 | 64 | |
AF428313.1 | - | 4 - 5 | 624 | 64 | ||
AY050375.1 | - | 4 - 5 | 722 | 64 | ||
BT009670.1 | - | 3 - 4 | 519 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |