Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGLG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337352.1 | NP_001325948.1 | 2 - 3 | 1305 | 117 | |
NM_111031.4 | NP_186814.1 | 2 - 3 | 807 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |