Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BET10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337357.1 | NP_001326912.1 | 5 - 6 | 1591 | 87 | |
NM_111043.3 | NP_566151.1 | 5 - 6 | 1602 | 87 | ||
GenBank mRNA | AY057662.1 | - | 5 - 6 | 1540 | 87 | |
AY099729.1 | - | 5 - 6 | 1547 | 87 | ||
AY128906.1 | - | 4 - 5 | 1292 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |