Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPLATE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111044.4 | NP_186827.2 | 5 - 6 | 2807 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |