Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337368.1 | NP_001319446.1 | 1 - 2 | 168 | 66 | |
NM_001337369.1 | NP_001327460.1 | 1 - 2 | 168 | 66 | ||
NM_001337370.1 | NP_001327455.1 | 1 - 2 | 91 | 66 | ||
NM_001337371.1 | NP_001327454.1 | 1 - 2 | 91 | 66 | ||
NM_001337372.1 | NP_001327459.1 | 1 - 2 | 91 | 66 | ||
NM_001337373.1 | NP_001327458.1 | 1 - 2 | 91 | 66 | ||
NM_001337374.1 | NP_001327457.1 | 1 - 2 | 91 | 66 | ||
NM_001337375.1 | NP_001327456.1 | 1 - 2 | 91 | 66 | ||
NM_001337376.1 | NP_001327453.1 | 1 - 2 | 168 | 66 | ||
NM_001337377.1 | NP_001327452.1 | 1 - 2 | 168 | 66 | ||
NM_111065.3 | NP_186848.1 | 1 - 2 | 168 | 66 | ||
GenBank mRNA | AF528062.1 | - | 1 - 2 | 168 | 66 | |
AK118222.1 | - | 1 - 2 | 120 | 66 | ||
BT005556.1 | - | 1 - 2 | 49 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |