Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KUP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111071.3 | NP_186854.1 | 1 - 2 | 252 | 72 | |
GenBank mRNA | AY062747.1 | - | 1 - 2 | 249 | 72 | |
BT003375.1 | - | 1 - 2 | 50 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |