Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111077.4 | NP_186860.1 | 2 - 3 | 641 | 72 | |
GenBank mRNA | AY037210.1 | - | 2 - 3 | 528 | 72 | |
BT002657.1 | - | 2 - 3 | 473 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |