Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111077.4 | NP_186860.1 | 7 - 8 | 1444 | 124 | |
GenBank mRNA | AY037210.1 | - | 7 - 8 | 1331 | 124 | |
BT002657.1 | - | 7 - 8 | 1276 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |