Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111088.4 | NP_186870.1 | 4 - 5 | 1182 | 85 | |
GenBank mRNA | AK118555.1 | - | 4 - 5 | 852 | 85 | |
BT005373.1 | - | 4 - 5 | 787 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |