Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337437.1 | NP_001325630.1 | 4 - 5 | 1131 | 95 | |
NM_001337438.1 | NP_001325629.1 | 4 - 5 | 950 | 95 | ||
NM_111153.4 | NP_186934.1 | 4 - 5 | 1131 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |