Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035549.1 | NP_001030626.1 | 5 - 6 | 470 | 66 | |
NM_001125086.1 | NP_001118558.1 | 5 - 6 | 461 | 66 | ||
NM_111155.3 | NP_186936.1 | 5 - 6 | 550 | 66 | ||
GenBank mRNA | AY035150.1 | - | 5 - 6 | 395 | 66 | |
AY063021.1 | - | 5 - 6 | 307 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |