Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VTC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125086.1 | NP_001118558.1 | 7 - 8 | 603 | 80 | |
NM_111155.3 | NP_186936.1 | 7 - 8 | 692 | 80 | ||
GenBank mRNA | AY035150.1 | - | 7 - 8 | 537 | 80 | |
AY063021.1 | - | 7 - 8 | 449 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |