Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111179.4 | NP_186959.2 | 4 - 5 | 539 | 77 | |
GenBank mRNA | AY140054.1 | - | 4 - 5 | 428 | 77 | |
BT010375.1 | - | 4 - 5 | 290 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |