Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111179.4 | NP_186959.2 | 7 - 8 | 829 | 99 | |
GenBank mRNA | AY140054.1 | - | 7 - 8 | 718 | 99 | |
BT010375.1 | - | 7 - 8 | 580 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |