Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111195.4 | NP_186975.1 | 18 - 19 | 1527 | 109 | |
GenBank mRNA | AY035071.1 | - | 18 - 19 | 1466 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |