Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337461.1 | NP_001327658.1 | 2 - 3 | 259 | 83 | |
NM_001337462.1 | NP_001327659.1 | 2 - 3 | 258 | 83 | ||
NM_111195.4 | NP_186975.1 | 2 - 3 | 259 | 83 | ||
GenBank mRNA | AY035071.1 | - | 2 - 3 | 198 | 83 | |
AY059148.1 | - | 2 - 3 | 112 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |