Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337463.1 | NP_001326716.1 | 2 - 3 | 284 | 116 | |
NM_111196.2 | NP_186976.2 | 2 - 3 | 284 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |