Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084632.1 | NP_001078101.1 | 8 - 9 | 1103 | 154 | |
NM_001202869.2 | NP_001189798.1 | 11 - 12 | 1466 | 154 | ||
NM_001337464.1 | NP_001327050.1 | 5 - 6 | 584 | 154 | ||
NM_111200.5 | NP_566199.4 | 12 - 13 | 1708 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |