Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084632.1 | NP_001078101.1 | 15 - 16 | 2717 | 67 | |
NM_001202869.2 | NP_001189798.1 | 18 - 19 | 3080 | 67 | ||
NM_001337464.1 | NP_001327050.1 | 12 - 13 | 2198 | 67 | ||
NM_111200.5 | NP_566199.4 | 19 - 20 | 3322 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |