Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337465.1 | NP_001327217.1 | 11 - 12 | 1197 | 151 | |
NM_111205.4 | NP_566203.2 | 11 - 12 | 1082 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |