Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337465.1 | NP_001327217.1 | 1 - 2 | 380 | 66 | |
NM_001337466.1 | NP_001327218.1 | 1 - 2 | 265 | 66 | ||
NM_111205.4 | NP_566203.2 | 1 - 2 | 265 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |