Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337469.1 | NP_001326670.1 | 7 - 8 | 1027 | 83 | |
NM_111209.3 | NP_566204.2 | 7 - 8 | 1027 | 83 | ||
GenBank mRNA | AY078951.1 | - | 7 - 8 | 912 | 83 | |
BT002727.1 | - | 7 - 8 | 807 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |