Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337481.1 | NP_001326505.1 | 1 - 2 | 675 | 117 | |
NM_111240.2 | NP_187019.1 | 1 - 2 | 528 | 117 | ||
GenBank mRNA | KJ138842.1 | - | 1 - 2 | 494 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |